Learn More
MP Biomedicals™ 3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™
Reagent that is excellent for ELISA procedures/immunoassays.
Brand: MP Biomedicals™ 08980501
Additional Details : CAS Number : 54827-17-7 Weight : 0.00500kg
Description
Molecular Formula: C16H20N2Molecular Weight: 240.3
Specifications
5 g | |
Powder | |
ELISA | |
Store at room temperature. Protect from light. |