missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog Number p-4812655
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
368-48-9
C6H4F3N
147.10
MFCD02183562
ATRQECRSCHYSNP-UHFFFAOYSA-N
2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine
136199
2-(trifluoromethyl)pyridine
FC(F)(F)C1=CC=CC=N1
This item is not returnable. View return policy
368-48-9
C6H4F3N
147.10
MFCD02183562
ATRQECRSCHYSNP-UHFFFAOYSA-N
2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine
136199
2-(trifluoromethyl)pyridine
FC(F)(F)C1=CC=CC=N1
Boiling Point 138°C to 141°C
Flash Point 42°C
Assay Percent Range 96% min. (GC)
Refractive Index 1.4183
Quantity 1 g
Formula Weight 147.1
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyridine
compliance-icons compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)pyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'