missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyrimidine-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3586681
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
122773-97-1
C11H8N2O2
200.197
MFCD00559612
BOAIYSRFGWBZCF-UHFFFAOYSA-N
2-phenyl-5-pyrimidinecarboxylic acid, 5-carboxy-2-phenylpyrimidine, 5-pyrimidinecarboxylic acid, 2-phenyl, 2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-phenyl, acmc-1c8my, 5-carboxy-2-phenyl-1,3-diazine, 2-phenyl pyrimidine-5-carboxylic acid
747389
2-phenylpyrimidine-5-carboxylic acid
C1=CC=C(C=C1)C2=NC=C(C=N2)C(=O)O
This item is not returnable. View return policy
122773-97-1
C11H8N2O2
200.197
MFCD00559612
BOAIYSRFGWBZCF-UHFFFAOYSA-N
2-phenyl-5-pyrimidinecarboxylic acid, 5-carboxy-2-phenylpyrimidine, 5-pyrimidinecarboxylic acid, 2-phenyl, 2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-phenyl, acmc-1c8my, 5-carboxy-2-phenyl-1,3-diazine, 2-phenyl pyrimidine-5-carboxylic acid
747389
2-phenylpyrimidine-5-carboxylic acid
C1=CC=C(C=C1)C2=NC=C(C=N2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.2
Percent Purity 97%
Chemical Name or Material 2-Phenylpyrimidine-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'