missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-N-Butoxyethanol, Extra Pure, SLR, Fisher Chemical™
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
2.5 L
25 L
Packaging:
Metal can
Plastic Coated Glass bottle
Description
2-N-Butoxyethanol, Extra Pure, SLR
Specifications
Melting Point | -70°C |
Color | Yellow |
Boiling Point | 171°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 500 mL |
Formula Weight | 118.18g/mol |
Vapor Pressure | 0.8 hPa at 20°C |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'