Learn More
2-Methylpyrimidine-5-carboxylic acid, 95%, Thermo Scientific™
Catalog Number p-7077482
Click to view available options
Quantity:
250 mg
1 g
5194-32-1
C6H6N2O2
138.13
NMGIXZFBQPETOK-UHFFFAOYSA-N
2-methyl-5-pyrimidinecarboxylic acid, 2-methylpyrimidine-5-carboxylicacid, 5-pyrimidinecarboxylic acid, 2-methyl, 2-methyl-5-pyrimidinecarboxylicacid, pubchem13290, acmc-209kwu, ksc269i7t, 2-methyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl, 2-methyl pyrimidine-5-carboxylic acid
13670875
2-methylpyrimidine-5-carboxylic acid
CC1=NC=C(C=N1)C(=O)O
5194-32-1
C6H6N2O2
138.13
NMGIXZFBQPETOK-UHFFFAOYSA-N
2-methyl-5-pyrimidinecarboxylic acid, 2-methylpyrimidine-5-carboxylicacid, 5-pyrimidinecarboxylic acid, 2-methyl, 2-methyl-5-pyrimidinecarboxylicacid, pubchem13290, acmc-209kwu, ksc269i7t, 2-methyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl, 2-methyl pyrimidine-5-carboxylic acid
13670875
2-methylpyrimidine-5-carboxylic acid
CC1=NC=C(C=N1)C(=O)O
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 2-Methylpyrimidine-5-carboxylic acid
compliance-icons
Product Identifier
  • 2-Methylpyrimidine-5-carboxylic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.