missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyridine-4-boronic acid, 95%, Thermo Scientific™

Catalog Number 15259926
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
579476-63-4
C6H8BNO2
136.95
MFCD03095364
UFYBTLOLWSABAU-UHFFFAOYSA-N
2-methylpyridine-4-boronic acid, 2-methylpyridin-4-yl boronic acid, 2-picoline-4-boronic acid, 2-methylpyridine-4-boronicacid, 4-borono-2-methylpyridine, 2-methyl-4-pyridineboronic acid, 2-methylpyridin-4-yl-4-boronic acid, 2-methyl-4-pyridinyl boronic acid, 2-methyl-4-pyridinyl-boronic acid, boronic acid, 2-methyl-4-pyridinyl
4192664
(2-methylpyridin-4-yl)boronic acid
CC1=CC(=CC=N1)B(O)O
This item is not returnable. View return policy
579476-63-4
C6H8BNO2
136.95
MFCD03095364
UFYBTLOLWSABAU-UHFFFAOYSA-N
2-methylpyridine-4-boronic acid, 2-methylpyridin-4-yl boronic acid, 2-picoline-4-boronic acid, 2-methylpyridine-4-boronicacid, 4-borono-2-methylpyridine, 2-methyl-4-pyridineboronic acid, 2-methylpyridin-4-yl-4-boronic acid, 2-methyl-4-pyridinyl boronic acid, 2-methyl-4-pyridinyl-boronic acid, boronic acid, 2-methyl-4-pyridinyl
4192664
(2-methylpyridin-4-yl)boronic acid
CC1=CC(=CC=N1)B(O)O
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 136.95
Percent Purity 95%
Chemical Name or Material 2-Methylpyridine-4-boronic acid
compliance-icons
Product Identifier
  • 2-Methylpyridine-4-boronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'