missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N,N-dimethylbenzamide, 97%

Brand:  Alfa Aesar™ H59416.03

Additional Details : CAS Number : 682757-75-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445728

  • 720.00 EUR / 1g


Estimated Shipment date
from Supplier 15-11-2019
Add to basket

Specifications

Chemical Identifiers

682757-75-1
182.198
OOOQACURFPURSF-UHFFFAOYSA-N
43510770
CN(C)C(=O)C1=C(C=CC(=C1)N)F
C9H11FN2O
MFCD01214099
5-amino-n,n-dimethyl-2-fluorobenzamide
5-amino-2-fluoro-N,N-dimethylbenzamide

Specifications

1g
Colorless
C9H11FN2O
5-amino-n,n-dimethyl-2-fluorobenzamide
CN(C)C(=O)C1=C(C=CC(=C1)N)F
182.198
167.18
97%
2-Fluoro-N,N-dimethylbenzamide
682757-75-1
MFCD01214099
OOOQACURFPURSF-UHFFFAOYSA-N
5-amino-2-fluoro-N,N-dimethylbenzamide
43510770
Liquid
Videos
SDS Documents