Learn More
Alfa Aesar™ 2-Fluoro-N-(4-fluoro-3-methylphenyl)benzamide, 97%
Brand:  Alfa Aesar™ H59638.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15446028
  • 720.00 EUR / 1g
Estimated Shipment date
from Supplier 20-06-2019
Add to basket
Specifications
Chemical Identifiers
294849-23-3
247.25
LLYFYFVGGFKIIW-UHFFFAOYSA-N
677523
C14H11F2NO
MFCD02169785
2-fluoro-n-4-fluoro-3-methylphenyl benzamide, 2-fluoro-n-4-fluoro-3-methylphenyl benzenecarboxamide
Specifications
2-Fluoro-N-(4-fluoro-3-methylphenyl)benzamide
294849-23-3
MFCD02169785
LLYFYFVGGFKIIW-UHFFFAOYSA-N
677523
Solid
1g
C14H11F2NO
2-fluoro-n-4-fluoro-3-methylphenyl benzamide, 2-fluoro-n-4-fluoro-3-methylphenyl benzenecarboxamide
247.25
247.24
97%