Learn More
IBA Solutions For Life Science 2'-Fluoro-2'-UTP
A modified nucleoside-triphosphate (modified 2´-ribonucleoside-5´-triphosphate) that prevents nuclease degradation of RNAs.
Brand:  IBA Solutions For Life Science 5-0611-774
Additional Details : Weight : 0.00050kg
Product Code. 15663628
  • 210.00 EUR / 1µmol
Estimated Shipment
03-09-2019
Add to basket
Description
Description
IBA has a nucleoside-triphosphate synthesis facility that offers an extensive range of triphosphates for many applications.
  • All products are double HPLC purified, desalted and well characterized by HPLC and 31P-NMR
  • They are also available as monophosphates and nucleosides
Specifications
Specifications
Yes
Prevents nuclease degradation of RNAs
1.0 μmol
Yes
Nucleoside-triphosphate