missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-1-methylpyridinium p-toluenesulfonate, tech. 90%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
2-Fluoro-1-methylpyridinium p-toluenesulfonate Is a peptide coupling reagent. Also used in the synthesis of ? -ribonucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-1-methylpyridinium p-toluenesulfonate Is a peptide coupling reagent. Also used in the synthesis of ɑ -ribonucleotides.
Solubility
Soluble in water (hydrolyzes), DMSO, dichloromethane, and acetonitrile.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Fluoro-1-methylpyridinium p-toluenesulfonate Is a peptide coupling reagent. Also used in the synthesis of ɑ -ribonucleotides.
Solubility
Soluble in water (hydrolyzes), DMSO, dichloromethane, and acetonitrile.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 128°C to 131°C |
| Quantity | 1 g |
| Beilstein | 4345357 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water (hydrolyzes),DMSO,dichloromethane,and acetonitrile. |
| Formula Weight | 283.32 |
| Percent Purity | 90% |
| Grade | Technical |
| Chemical Name or Material | 2-Fluoro-1-methylpyridinium p-toluenesulfonate |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction