Learn More
Thermo Scientific Chemicals 2'-Deoxythymidine-5'-diphosphate trisodium salt, 97%
Click to view available options
Quantity:
25 mg
100 mg
Description
2'-Deoxythymidine-5'-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′-Deoxythymidine-5′-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2′-Deoxythymidine-5′-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Color | White |
Quantity | 25 mg |
Solubility Information | Soluble in water. |
Formula Weight | 468.13 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | 2'-Deoxythymidine-5'-diphosphate trisodium salt |