missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloropyridine-4-boronic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 458532-96-2 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C5H5BClNO2 |
MDL Number | MFCD03788416 |
Quantity | 1 g |
Synonym | 2-chloro-4-pyridylboronic acid, 2-chloropyridine-4-boronic acid, 2-chloropyridin-4-yl boronic acid, 2-chloro-4-pyridineboronic acid, 2-chloropyridin-4-yl-4-boronic acid, 2-chloropyridine-4-boronoic acid, 2-chloropyridin-4-boronic acid, 2-chloropyridine-4-boronicacid, boronic acid, 2-chloro-4-pyridinyl, 2-chloro-4-pyridyl boronic acid |
InChI Key | WJYRVVDXJMJLTN-UHFFFAOYSA-N |
SMILES | B(C1=CC(=NC=C1)Cl)(O)O |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'