Learn More
Alfa Aesar™ 2-Chloro-N-(2,4-dimethoxyphenyl)benzamide, 97%
Brand:  Alfa Aesar™ H59226.03
Additional Details : CAS Number : 196938-29-1 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15485468
  • 646.00 EUR / 1g
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Chemical Identifiers
196938-29-1
291.731
SINZCIGNCGXPIB-UHFFFAOYSA-N
C15H14ClNO3
MFCD00032669
722324
Specifications
2-Chloro-N-(2,4-dimethoxyphenyl)benzamide
196938-29-1
Solid
C15H14ClNO3
SINZCIGNCGXPIB-UHFFFAOYSA-N
2-chloro-N-(2,4-dimethoxyphenyl)benzamide
722324
291.73
1g
97%
MFCD00032669
COC1=CC(=C(C=C1)NC(=O)C2=CC=CC=C2Cl)OC
291.731