missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
Specifications
Melting Point | 43°C |
Quantity | 250 mg |
Beilstein | 10249610 |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 307.44 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'