Learn More
Alfa Aesar™ 2-Chloro-4-nitro-N-[4-(trifluoromethyl)phenyl]benzamide, 97%
Brand:  Alfa Aesar™ H59619.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15485136
  • 646.00 EUR / 1g
Estimated Shipment date
from Supplier 31-01-2018
Add to basket
Specifications
Chemical Identifiers
2-Chloro-4-nitro-N-[4-(trifluoromethyl)phenyl]benzamide
344.67
MFCD04554330
Specifications
2-Chloro-4-nitro-N-[4-(trifluoromethyl)phenyl]benzamide
Solid
344.67
1g
97%
MFCD04554330