missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzoic acid, 99%, Thermo Scientific™

Catalog Number p-87271
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2252-51-9
C7H4ClFO2
174.56
MFCD00010615
GRPWQLDSGNZEQE-UHFFFAOYSA-N
benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
75259
2-chloro-4-fluorobenzoic acid
C1=CC(=C(C=C1F)Cl)C(=O)O
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2252-51-9
C7H4ClFO2
174.56
MFCD00010615
GRPWQLDSGNZEQE-UHFFFAOYSA-N
benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
75259
2-chloro-4-fluorobenzoic acid
C1=CC(=C(C=C1F)Cl)C(=O)O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 183°C to 186°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(F)CO2H
Quantity 5 g
Formula Weight 174.56
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material 2-Chloro-4-fluorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'