missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-72721
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
88-65-3
C7H5BrO2
201.02
MFCD00002402
XRXMNWGCKISMOH-UHFFFAOYSA-N
o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657
6940
2-bromobenzoic acid
C1=CC=C(C(=C1)C(=O)O)Br
This item is not returnable. View return policy
88-65-3
C7H5BrO2
201.02
MFCD00002402
XRXMNWGCKISMOH-UHFFFAOYSA-N
o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657
6940
2-bromobenzoic acid
C1=CC=C(C(=C1)C(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 146.0°C to 151.0°C
Color Beige or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4CO2H
Quantity 25 g
Beilstein 09, 347
Solubility Information Solubility in water: slightly soluble
Formula Weight 201.02
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Bromobenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'