missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine, 98%

Brand:  Alfa Aesar™ H63801.03

Additional Details : CAS Number : 49669-14-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458526

  • 45.00 EUR / 1g


Estimated Shipment date
from Supplier 18-06-2019
Add to basket

Specifications

Chemical Identifiers

49669-14-9
244.088
ZRXWMMNBTGBCIL-UHFFFAOYSA-N
12164282
CC1(OCCO1)C2=NC(=CC=C2)Br
C9H10BrNO2
MFCD12406888
2-bromo-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl-pyridine, pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl, pubchem16082, ksc496i5p, 2-bromo-6-2-methyl-1,3-dioxolane-2-yl pyridine, 2-bromanyl-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3 dioxolan-2-yl-pyridine, 6-bromo-2-2'-methyl-1',3'-dioxolan-2'-yl-pyridine
2-bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine

Specifications

2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
49669-14-9
MFCD12406888
ZRXWMMNBTGBCIL-UHFFFAOYSA-N
2-bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
12164282
Solid
1g
C9H10BrNO2
2-bromo-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl-pyridine, pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl, pubchem16082, ksc496i5p, 2-bromo-6-2-methyl-1,3-dioxolane-2-yl pyridine, 2-bromanyl-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3 dioxolan-2-yl-pyridine, 6-bromo-2-2'-methyl-1',3'-dioxolan-2'-yl-pyridine
CC1(OCCO1)C2=NC(=CC=C2)Br
244.088
244.09
98%
Videos
SDS