Learn More
Alfa Aesar™ 2-(Boc-amino)-4-fluorobenzeneboronic acid pinacol ester, 96%
Brand:  Alfa Aesar™ H62383.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15487356
  • 685.00 EUR / 1g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
2-(Boc-amino)-4-fluorobenzeneboronic acid pinacol ester
337.19
2-(Boc-amino)-4-fluorophenylboronic acid pinacol ester
Specifications
2-(Boc-amino)-4-fluorobenzeneboronic acid pinacol ester
96%
1g
Solid
337.19
2-(Boc-amino)-4-fluorophenylboronic acid pinacol ester