missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzyl-4-chlorophenol, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Used in soaps, anionic detergents, cosmetics and aerosol spray products. It is a useful biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Benzyl-4-chlorophenol is used in formulating disinfectant and sanitizer products. It is also used in soaps, anionic detergents, cosmetics and aerosol spray products. It is a useful biochemical for proteomics research.
Solubility
Soluble in ethanol, alkaline solutions. Soluble in water(<1mg/mL).
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
2-Benzyl-4-chlorophenol is used in formulating disinfectant and sanitizer products. It is also used in soaps, anionic detergents, cosmetics and aerosol spray products. It is a useful biochemical for proteomics research.
Solubility
Soluble in ethanol, alkaline solutions. Soluble in water(<1mg/mL).
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Specifications
Melting Point | 45°C to 51°C |
Density | 1.19 |
Boiling Point | 160°C to 162°C (5 mmHg) |
Flash Point | 185°C (365°F) |
Quantity | 25 g |
UN Number | UN3077 |
Beilstein | 1959194 |
Merck Index | 14,2407 |
Solubility Information | Soluble in ethanol,alkaline solutions. Soluble in water(<1mg/ml). |
Formula Weight | 218.69 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'