missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 235.0°C to 238.0°C |
Boiling Point | 314.0°C |
Infrared Spectrum | Authentic |
Quantity | 5 g |
Solubility Information | Solubility in water: slightly soluble. |
Formula Weight | 129.55 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-5-chloropyrimidine |
Safety and Handling
Product Identifier
- 2-Amino-5-chloropyrimidine
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'