missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-3756771
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
719-59-5
C13H10ClNO
231.68
MFCD00007839
ZUWXHHBROGLWNH-UHFFFAOYSA-N
2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone
12870
(2-amino-5-chlorophenyl)-phenylmethanone
NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
719-59-5
C13H10ClNO
231.68
MFCD00007839
ZUWXHHBROGLWNH-UHFFFAOYSA-N
2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone
12870
(2-amino-5-chlorophenyl)-phenylmethanone
NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 96°C to 100°C
Color Yellow
Boiling Point 207°C
Flash Point 211°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula H2NC6H3(Cl)COC6H5
Quantity 25 g
Beilstein 14, 79
Solubility Information Solubility in water: practically insoluble
Formula Weight 231.68
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chlorobenzophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-chlorobenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'