missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxamide, 97%, Thermo Scientific™

Catalog Number p-4812735
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4815-28-5
C9H12N2OS
196.27
FFAKFORHXDNYEN-UHFFFAOYSA-N
2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide, benzo b thiophene-3-carboxamide, 2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide, 2-amino-3-carboxamido-4,5,6,7-tetrahydro benzo b thiophene, benzo b thiophene-3-carboxamide,2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxamide, 2-amino-3-carbamoyl-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-3-carboxamido-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-4,5,6,7-tetrahydro-benzo b thiophen-3-carboxamid, 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carboxamide
290223
2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
C1CCC2=C(C1)C(=C(S2)N)C(=O)N
This item is not returnable. View return policy
4815-28-5
C9H12N2OS
196.27
FFAKFORHXDNYEN-UHFFFAOYSA-N
2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide, benzo b thiophene-3-carboxamide, 2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide, 2-amino-3-carboxamido-4,5,6,7-tetrahydro benzo b thiophene, benzo b thiophene-3-carboxamide,2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxamide, 2-amino-3-carbamoyl-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-3-carboxamido-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-4,5,6,7-tetrahydro-benzo b thiophen-3-carboxamid, 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carboxamide
290223
2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
C1CCC2=C(C1)C(=C(S2)N)C(=O)N
Melting Point 181°C to 183°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxamide
compliance-icons
Product Identifier
  • 2-Amino-4,5,6,7-tetrahydro-1-benzo¢b!thiophene-3-carboxamide, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'