missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-methoxybenzylamine, 97%, Thermo Scientific™

Catalog Number 11451400
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
771573-20-7
C8H9F2NO
173.16
MFCD04115924
ALINWWVXTNYKIM-UHFFFAOYSA-N
2,6-difluoro-4-methoxybenzylamine, 2,6-difluoro-4-methoxyphenyl methanamine, 2,6-difluoro-4-methoxy-phenyl methanamine, 1-2,6-difluoro-4-methoxyphenyl methanamine, 2,6-difluoro-4-methoxyphenyl methylamine, 2,6-bis fluoranyl-4-methoxy-phenyl methanamine, benzenemethanamine, 2,6-difluoro-4-methoxy-9ci
3866087
(2,6-difluoro-4-methoxyphenyl)methanamine
COC1=CC(F)=C(CN)C(F)=C1
This item is not returnable. View return policy
771573-20-7
C8H9F2NO
173.16
MFCD04115924
ALINWWVXTNYKIM-UHFFFAOYSA-N
2,6-difluoro-4-methoxybenzylamine, 2,6-difluoro-4-methoxyphenyl methanamine, 2,6-difluoro-4-methoxy-phenyl methanamine, 1-2,6-difluoro-4-methoxyphenyl methanamine, 2,6-difluoro-4-methoxyphenyl methylamine, 2,6-bis fluoranyl-4-methoxy-phenyl methanamine, benzenemethanamine, 2,6-difluoro-4-methoxy-9ci
3866087
(2,6-difluoro-4-methoxyphenyl)methanamine
COC1=CC(F)=C(CN)C(F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.5055
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 173.16
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-4-methoxybenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'