missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(5-Methyl-2-furyl)-5-nitrobenzimidazole, 95%

Brand:  Alfa Aesar™ H64090.MD

Additional Details : CAS Number : 37128-68-0 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15458696

  • 108.00 EUR / 250mg


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

37128-68-0
243.222
JSCVVMSVSAVEJN-UHFFFAOYSA-N
1088924
CC1=CC=C(O1)C2=NC3=C(N2)C=C(C=C3)[N+](=O)[O-]
C12H9N3O3
MFCD03001005
2-5-methyl-furan-2-yl-5-nitro-1h-benzoimidazole, 2-5-methyl-2-furyl-5-nitrobenzimidazole, 2-5-methylfuran-2-yl-6-nitro-1h-benzimidazole, 2-5-methylfuran-2-yl-5-nitro-1h-1,3-benzodiazole
2-(5-methylfuran-2-yl)-6-nitro-1H-benzimidazole

Specifications

2-(5-Methyl-2-furyl)-5-nitrobenzimidazole
37128-68-0
MFCD03001005
JSCVVMSVSAVEJN-UHFFFAOYSA-N
2-(5-methylfuran-2-yl)-6-nitro-1H-benzimidazole
1088924
Solid
250mg
C12H9N3O3
2-5-methyl-furan-2-yl-5-nitro-1h-benzoimidazole, 2-5-methyl-2-furyl-5-nitrobenzimidazole, 2-5-methylfuran-2-yl-6-nitro-1h-benzimidazole, 2-5-methylfuran-2-yl-5-nitro-1h-1,3-benzodiazole
CC1=CC=C(O1)C2=NC3=C(N2)C=C(C=C3)[N+](=O)[O-]
243.222
243.22
95%
Videos
SDS