Learn More
Alfa Aesar™ 2-(5-Bromo-2-furyl)thiazolidine, 97%
Brand:  Alfa Aesar™ H62897.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15459348
  • 257.00 EUR / 1g
Estimated Shipment date
from Supplier 19-02-2019
Add to basket
Specifications
Chemical Identifiers
MFCD16685164
5-Bromo-2-(2-thiazolidinyl)furan
Specifications
2-(5-Bromo-2-furyl)thiazolidine
MFCD16685164
234.11
97%
1g
5-Bromo-2-(2-thiazolidinyl)furan
Solid