missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dimethoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65357.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459586

  • 256.00 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD01232846

Specifications

2,4-Dimethoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
MFCD01232846
Solid
1g
311.38
95%
Videos
SDS