missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorobenzyl alcohol, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used to produce 1-bromomethyl-2,3-difluoro-benzene with reagent PBr3 and solvent toluene at the temperature of 100°C for 1 hour. 2,3-Difluorobenzyl alcohol may be used in chemical synthesis studies.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used to produce 1-bromomethyl-2,3-difluoro-benzene with reagent PBr3 and solvent toluene at the temperature of 100°C for 1 hour. 2,3-Difluorobenzyl alcohol may be used in chemical synthesis studies.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Density | 1.282 |
Flash Point | 93°C (199°F) |
Refractive Index | 1.492 |
Quantity | 5 g |
Beilstein | 7089243 |
Synonym | 2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol |
Solubility Information | Slightly soluble in water. |
Formula Weight | 144.12 |
Percent Purity | 97% |
Chemical Name or Material | 2,3-Difluorobenzyl alcohol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'