missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzyl alcohol, 97%, Thermo Scientific™

Catalog Number 11430057
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
75853-18-8
C7H6F2O
144.121
MFCD00010312
JSFGDUIJQWWBGY-UHFFFAOYSA-N
447153
CHEBI:41883
(2,3-difluorophenyl)methanol
C1=CC(=C(C(=C1)F)F)CO
This item is not returnable. View return policy
75853-18-8
C7H6F2O
144.121
MFCD00010312
JSFGDUIJQWWBGY-UHFFFAOYSA-N
447153
CHEBI:41883
(2,3-difluorophenyl)methanol
C1=CC(=C(C(=C1)F)F)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used to produce 1-bromomethyl-2,3-difluoro-benzene with reagent PBr3 and solvent toluene at the temperature of 100°C for 1 hour. 2,3-Difluorobenzyl alcohol may be used in chemical synthesis studies.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 1.282
Flash Point 93°C (199°F)
Refractive Index 1.492
Quantity 5 g
Beilstein 7089243
Synonym 2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol
Solubility Information Slightly soluble in water.
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 2,3-Difluorobenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'