missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorobenzyl alcohol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
25 g
Specifications
Density | 1.2800g/mL |
Flash Point | 93°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | F2C6H3CH2OH |
Refractive Index | 1.4910 to 1.4930 |
Quantity | 1 g |
Specific Gravity | 1.28 |
Formula Weight | 144.12 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'