missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(5-fluoro-2-methylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65331.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459566

  • 285.00 EUR / 1g


Estimated Shipment date
from Supplier 18-06-2019
Add to basket

Specifications

Chemical Identifiers

MFCD28098785

Specifications

2-(3-Benzoylphenyl)-N-(5-fluoro-2-methylphenyl)propionamide
MFCD28098785
Solid
1g
361.41
95%
Videos
SDS