missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-methoxybenzyl)propionamide, 95%

Brand:  Alfa Aesar™ H65306.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459556

  • 256.00 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD28098783

Specifications

2-(3-Benzoylphenyl)-N-(4-methoxybenzyl)propionamide
MFCD28098783
Solid
1g
373.44
95%
Videos
SDS