missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorostyrene, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number p-178727
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
653-34-9, 123-31-9
C8H3F5
194.1
MFCD00000300
LVJZCPNIJXVIAT-UHFFFAOYSA-N
2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
69556
1-ethenyl-2,3,4,5,6-pentafluorobenzene
C=CC1=C(C(=C(C(=C1F)F)F)F)F
This item is not returnable. View return policy
653-34-9, 123-31-9
C8H3F5
194.1
MFCD00000300
LVJZCPNIJXVIAT-UHFFFAOYSA-N
2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
69556
1-ethenyl-2,3,4,5,6-pentafluorobenzene
C=CC1=C(C(=C(C(=C1F)F)F)F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4060g/mL
Boiling Point 62.0°C to 63.0°C (50.0 mmHg)
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6F5CH=CH2
Refractive Index 1.4445 to 1.4465
Quantity 5 g
Specific Gravity 1.406
Stabilizer 0.01% tert.-butylcatechol
Formula Weight 194.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3,4,5,6-Pentafluorostyrene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4,5,6-Pentafluorostyrene, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'