missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Thiodiethanol, for Amino Acid Analysis, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Plastic Coated Glass bottle
Specifications
CAS | 111-48-8 |
Boiling Point | 165°C |
Packaging | Plastic Coated Glass bottle |
Molecular Formula | C4H10O2S |
MDL Number | 2910 |
Quantity | 100 g |
Synonym | thiodiglycol, 2,2'-thiodiethanol, thiodiethanol, thiodiethylene glycol, tedegyl, kromfax solvent, bis 2-hydroxyethyl sulfide, ethanol, 2,2'-thiobis, 2,2'-thiobisethanol |
InChI Key | YODZTKMDCQEPHD-UHFFFAOYSA-N |
SMILES | C(CSCCO)O |
IUPAC Name | 2-(2-hydroxyethylsulfanyl)ethanol |
Show More |
Safety and Handling
Product Identifier
- 2,2''-Thiodiethanol
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'