Learn More
Alfa Aesar™ 2-(2-Pyridyl)thiazolidine-4-carboxylic acid, 97%
Brand:  Alfa Aesar™ H62636.03
Additional Details : CAS Number : 84682-22-4 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15459138
  • 257.00 EUR / 1g
Estimated Shipment date
from Supplier 24-04-2019
Add to basket
Specifications
Chemical Identifiers
84682-22-4
210.251
FRABGDHEKOTVGC-UHFFFAOYSA-N
3020041
C1C(NC(S1)C2=CC=CC=N2)C(=O)O
C9H10N2O2S
MFCD09949861
2-pyridin-2-ylthiazolidine-4-carboxylic acid, 2-pyridin-2-yl-1,3-thiazolidine-4-carboxylic acid, acmc-20ltaq, acmc-20ltar, 4-thiazolidinecarboxylic acid, 2-2-pyridinyl-, 2r,4r, 4-thiazolidinecarboxylic acid, 2-2-pyridinyl-, 2s,4r, 2-2-pyridinyl-4-thiazolidinecarboxylic acid, pyridin-2-yl-1,3-thiazolidine-4-carboxylic acid
2-pyridin-2-yl-1,3-thiazolidine-4-carboxylic acid
Specifications
2-(2-Pyridyl)thiazolidine-4-carboxylic acid
84682-22-4
MFCD09949861
FRABGDHEKOTVGC-UHFFFAOYSA-N
2-pyridin-2-yl-1,3-thiazolidine-4-carboxylic acid
3020041
Solid
1g
C9H10N2O2S
2-pyridin-2-ylthiazolidine-4-carboxylic acid, 2-pyridin-2-yl-1,3-thiazolidine-4-carboxylic acid, acmc-20ltaq, acmc-20ltar, 4-thiazolidinecarboxylic acid, 2-2-pyridinyl-, 2r,4r, 4-thiazolidinecarboxylic acid, 2-2-pyridinyl-, 2s,4r, 2-2-pyridinyl-4-thiazolidinecarboxylic acid, pyridin-2-yl-1,3-thiazolidine-4-carboxylic acid
C1C(NC(S1)C2=CC=CC=N2)C(=O)O
210.251
210.25
97%