missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine, 98%, Thermo Scientific™

Catalog Number 15459166
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
152460-09-8
C16H13N5O2
307.313
MFCD02179269
OJITWRFPRCHSMX-UHFFFAOYSA-N
n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine, chembl20426, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine, n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine, 2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine, 2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine, 2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl, n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
10686149
N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3
This item is not returnable. View return policy
152460-09-8
C16H13N5O2
307.313
MFCD02179269
OJITWRFPRCHSMX-UHFFFAOYSA-N
n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine, chembl20426, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine, n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine, 2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine, 2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine, 2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl, n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
10686149
N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 188°C to 193°C
Color Yellow
Quantity 1 g
Beilstein 7544032
Formula Weight 307.31
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'