missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,6,6-Tetramethylpiperidinooxy, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 36°C to 40°C |
Color | Yellow to Green |
Flash Point | 67°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5 % min. (GC) |
Packaging | Glass bottle |
Quantity | 5 g |
Fieser | 16,324; 17,332 |
Show More |
Safety and Handling
Product Identifier
- 2,2,6,6-Tetramethylpiperidinooxy
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'