missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetramethoxybutane, 95%, Thermo Scientific Chemicals

Catalog Number 15294258
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
176798-33-7
C8H18O4
178.23
MFCD25458126
COEBCOIFCLAOFZ-UHFFFAOYSA-N
butane, 2,2,3,3-tetramethoxy
10856041
2,2,3,3-tetramethoxybutane
COC(C)(OC)C(C)(OC)OC
This item is not returnable. View return policy
176798-33-7
C8H18O4
178.23
MFCD25458126
COEBCOIFCLAOFZ-UHFFFAOYSA-N
butane, 2,2,3,3-tetramethoxy
10856041
2,2,3,3-tetramethoxybutane
COC(C)(OC)C(C)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 172.0°C to 174.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 178.23
Percent Purity 95%
Chemical Name or Material 2,2,3,3-Tetramethoxybutane
compliance-icons
Product Identifier
  • 2,2,3,3-Tetramethoxybutane, 95%
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'