missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals

Catalog Number p-3758338
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
108-65-6
C6H12O3
132.16
MFCD00038500
LLHKCFNBLRBOGN-UHFFFAOYSA-N
1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester
7946
1-methoxypropan-2-yl acetate
CC(COC)OC(=O)C
108-65-6
C6H12O3
132.16
MFCD00038500
LLHKCFNBLRBOGN-UHFFFAOYSA-N
1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester
7946
1-methoxypropan-2-yl acetate
CC(COC)OC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -87.0°C
Color Colorless
Density 0.9600g/mL
Boiling Point 146.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CH(CH3)OC(O)CH3
Refractive Index 1.4010 to 1.4030
Quantity 1 kg
UN Number 1993
Specific Gravity 0.96
Solubility Information Solubility in water: 19.8g/L (25°C)
Formula Weight 132.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(1-Methoxy)propyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • 2-(1-Methoxy)propyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'