missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.
Solubility
Slightly soluble in water.
Notes
Store away from acids, air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Keep it refrigerated.
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.
Solubility
Slightly soluble in water.
Notes
Store away from acids, air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Keep it refrigerated.
Specifications
Melting Point | 39°C to 44°C |
Color | Brown |
Density | 0.902 g/mL |
Flash Point | 74°C (165°F) |
Refractive Index | 1.472 |
Quantity | 5 g |
UN Number | UN3259 |
Beilstein | 4304824 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 142.25 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'