Learn More
Trajan Scientific and Medical™ 1MR-VLL-GT Pre-fitted Syringe Valve
Ideal for sample storage and transportation or to pressurize sample prior to injection.
Brand:  Trajan Scientific and Medical™ 008160
Additional Details : Weight : 0.06000kg
Product Code. 11932595
  • 180.00 EUR / Each
Estimated Shipment: 03-07-2024
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Pre-fitted with a valve for sample pressurization prior to injection
Specifications
Specifications
PTFE (Plunger)
Pre-fitted Syringe Valve
1 mL
For sample storage and transportation or to pressurize sample prior to injection
1MR-VLL-GT