Learn More
Sartorius™ 18406 Series Non-Sterile Regenerated Cellulose (RC) Membrane Filters
18406 Series Non-Sterile Regenerated Cellulose (RC) Membrane Filters are solvent-resistant and hydrophilic, with low non-specific adsorption. Reinforced with nonwoven cellulose, they are ideal for the clarification and degassing of solvents and mobile phases for HPLC.
176.00 EUR - 278.00 EUR
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
For Use With (Application) | Suited for their major application, particle removal from solvents |
Material (Membrane) | Regenerated Cellulose |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) Membrane | Pore Size | Bubble Point | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Pore Size | Bubble Point | Price | Quantity & Availability | |||
10253331
|
Sartorius™
18407-100------G |
100 mm | 0.2 μm | 4.4bar |
176.00 EUR
Pack of 25 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
11770696
|
Sartorius™
18406--50----ACN |
50 mm | 0.45 μm | 2.9bar |
278.00 EUR
Pack of 100 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
Description
- Product Type: Membrane Filter
- Material: Regenerated Cellulose
- Membrane Color: White
- Membrane Thickness: 150 μm
- Wettability: Hydrophilic
- Sterility: Non-Sterile
- For Use With (Application): Analytical Sample Preparation, Sterile Filtration, Filtration of Aqueous Solutions, Solvent Filtration, Clarification
Specifications
Autoclavable | |
Suited for their major application, particle removal from solvents | |
Non-sterile | |
Membrane Filter |
White | |
Regenerated Cellulose | |
170 μm |