Learn More
Bochem™ 18/10 Stainless Steel Chattaway Double Spatulas
Brand:  Bochem™ 3142
Additional Details : Weight : 0.01080kg
 View more versions of this product
Product Code. 10365811
  • 2.50 EUR / Each
Estimated Shipment: 12-06-2024
Specifications
1 Ea.
40 mm
7 mm
3.5mm
Stainless Steel (18/10)
Chattaway Double Spatula
7 mm
Stainless Steel (18/10)
150 x 7 mm
150 mm