Learn More
Hamilton™ Long Life Instrument Syringes
Long life syringes were developed for extreme conditions that include harsh chemicals as well as abrasive particulate matter.
Brand:  Hamilton™ 8250-20
Additional Details : Weight : 0.03800kg
 View more versions of this product
Product Code. 10034194
  • 212.00 EUR / Each
Estimated Shipment: 13-05-2024
Description
  • Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
  • Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications
Specifications
Borosilicate Glass, PTFE, UHMWPE
Instrument Syringe
50 μL
7.73 mm
1/Ea.
Luer Lock