missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ 11304 Series Non-Sterile Cellulose Nitrate (CN) Membrane Filters

11304 Series Non-Sterile Cellulose Nitrate (CN) Membrane Filters are hydrophilic, have high flow rates thanks to their symmetrical structure and are compatible with aqueous solutions (pH 4 – 8), hydrocarbons, and several other organic solvents.

Brand:  Sartorius™ 11304-100------G

Additional Details : Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10781553

  • 216.00 EUR / Pack of 25
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High, non-specific absorption
  • Hydrophilic, with high flow rates
  • Available in a range of pore sizes and diameters
  • Ideal for several laboratory applications including particle retention, cell capture, cell retention, and cell filtration
Specifications

Specifications

Autoclavable
White
Cellulose Nitrate
0.8 μm
200mL/min./cm2/bar
25/Pk
Membrane Filter
100 mm
1.4bar
Membrane Filter
Cellulose Nitrate
Non-sterile
For many general laboratory applications where a membrane with a high non-specific adsorption is suitable
130 μm
100 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P230-Keep wetted with .
  • P233-Keep container tightly closed.
  • P234-Keep only in original container.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use to extinguish.
  • P372-Explosion risk in case of fire.
  • P375-Fight fire remotely due to the risk of explosion.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers