Learn More
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%, Thermo Scientific Chemicals
Catalog Number 15495135
Click to view available options
Quantity:
1 g
5 g
207399-07-3
C36H44ClIN2
667.116
MFCD00191907
IRPKBYJYVJOQHQ-UHFFFAOYSA-M
2-2-2-chloro-3-1,3-dihydro-3,3-dimethyl-1-propyl-2h-indol-2-ylidene ethylidene-1-cyclohexen-1-yl ethenyl-3,3-dimethyl-1-propylindolium iodide
118987185
2-[(2E)-2-[2-chloro-3-[(E)-2-(3,3-dimethyl-1-propylindol-1-ium-2-yl)ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethyl-1-propylindole;iodide
CCCN1C2=CC=CC=C2C(C1=CC=C3CCCC(=C3Cl)C=CC4=[N+](C5=CC=CC=C5C4(C)C)CCC)(C)C.[I-]
207399-07-3
C36H44ClIN2
667.116
MFCD00191907
IRPKBYJYVJOQHQ-UHFFFAOYSA-M
2-2-2-chloro-3-1,3-dihydro-3,3-dimethyl-1-propyl-2h-indol-2-ylidene ethylidene-1-cyclohexen-1-yl ethenyl-3,3-dimethyl-1-propylindolium iodide
118987185
2-[(2E)-2-[2-chloro-3-[(E)-2-(3,3-dimethyl-1-propylindol-1-ium-2-yl)ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethyl-1-propylindole;iodide
CCCN1C2=CC=CC=C2C(C1=CC=C3CCCC(=C3Cl)C=CC4=[N+](C5=CC=CC=C5C4(C)C)CCC)(C)C.[I-]
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
11-Chloro-1,1′-di-n-propyl-3,3,3′,3′-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.
Solubility
It is soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 232°C to 234°C
Quantity 1 g
Solubility Information It is soluble in methanol.
Formula Weight 667.12
Percent Purity 95%
Chemical Name or Material 11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide