missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ischemin sodium salt

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ischemin sodium salt is a CBP bromodomain inhibitor. Inhibits p53 interaction with CBP and decreases p53 transcriptional activity. Protects against Doxorubicin -induced apoptosis in cardiomyocytes in vitro. Cell permeable.
Specifications
Specifications
| Purity | >99% |
| Quantity | 10 mg |
| Product Type | Ischemin sodium salt |
| Molecular Weight (g/mol) | 357.36 |
| Inhibitors | Ischemin sodium salt |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction