Learn More
100ML Iron 1000 µg/ml BAKER INSTRA-ANALYZED Reagent
Brand: JT Baker 5764.0100
100ML Iron 1000 µg/ml BAKER INSTRA-ANALYZED Reagent
Additional Details : Weight : 0.10000kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Safety and Handling
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising liquid Category 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.