missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ 1000mL Media Bottle, Clear Type I
Click to view available options
Closure Included:
Yes
Description
- Clear borosilicate glass bottles, non-graduated
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Hazardous communication label included for each case for content ID and safety notification
- Tamper evident seal keeps lidded bottles sealed to protect contents from accidental misuse
Specifications
Closure Liner | Polyethylene |
Closure Material | Phenolic |
Closure Size | 38-430 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 103 mm |
Height (Metric) | 231 mm |
Product Type | Media Bottle |
Capacity (Metric) | 1000 mL |
Color | Clear |
Includes | Pour lip |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'