missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthyl Phosphate Monosodium Salt Monohydrate, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Physical Form | Powder |
Quantity | 1 g |
Solubility Information | Soluble in water (50mg/mL) |
IUPAC Name | naphthalen-1-yl phosphate |
Formula Weight | 264.87g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'